Thermo Scientific Chemicals

4-Ethylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: 118950250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Ethylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: 118950250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118950250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2896-60-8
IUPAC Name
4-ethylbenzene-1,3-diol
Molecular Formula
C8H10O2
InChI Key
VGMJYYDKPUPTID-UHFFFAOYSA-N
SMILES
CCC1=CC=C(O)C=C1O
Appearance (Color)
Light beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
95°C to 99°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text