Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals
Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 119020250
Price (USD)/ Each
49.30
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.30
Add to cart
Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals
Catalog number119020250
Price (USD)/ Each
49.30
-
Add to cart
Chemical Identifiers
CAS383-63-1
IUPAC Nameethyl 2,2,2-trifluoroacetate
Molecular FormulaC4H5F3O2
InChI KeySTSCVKRWJPWALQ-UHFFFAOYSA-N
SMILESCCOC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only