Thermo Scientific Chemicals

Ethyl 4,4,4-trifluoroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 119030250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 4,4,4-trifluoroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: 119030250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 119030250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
372-31-6
IUPAC Name
ethyl 4,4,4-trifluoro-3-oxobutanoate
Molecular Formula
C6H7F3O3
InChI Key
OCJKUQIPRNZDTK-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)C(F)(F)F
Appearance (Form)
Liquid
GC
>=96.0 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.3750 to 1.3780 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text