Thermo Scientific Chemicals

Ethyltriphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: 119045000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyltriphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: 119045000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 119045000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1530-32-1
IUPAC Name
ethyltriphenylphosphanium bromide
Molecular Formula
C20H20BrP
InChI Key
JHYNXXDQQHTCHJ-UHFFFAOYSA-M
SMILES
[Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 % (on dry substance)
Appearance (Form)
Crystalline powder
Melting point
206.5°C to 208.5°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text