Eugenol, 99%
Eugenol, 99%
Eugenol, 99%
Thermo Scientific Chemicals

Eugenol, 99%

CAS: 97-53-0 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 119111000
Price (EUR)
30,26
Online Exclusive
35,60
Save 5,34 (15%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
30,26
Online Exclusive
35,60
Save 5,34 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name2-methoxy-4-(prop-2-en-1-yl)phenol
Molecular FormulaC10H12O2
InChI KeyRRAFCDWBNXTKKO-UHFFFAOYSA-N
SMILESCOC1=CC(CC=C)=CC=C1O
Molecular Weight (g/mol)164.20
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(20°C) 1.064 to 1.070
Appearance (Color)Clear colorless to yellow-green
Infrared spectrumConforms
Refractive index1.5400 to 1.5420 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link