Thermo Scientific Chemicals

Farnesol, 96%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 119120250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Farnesol, 96%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 119120250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 119120250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4602-84-0
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Density
(20/4°C) 0.884 to 0.89 kg/l
Infrared spectrum
Conforms
GC
>=95 % (sum of geometric isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text