Flavone, 99+%, Thermo Scientific Chemicals
Flavone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Flavone, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 119160250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O2
InChI KeyVHBFFQKBGNRLFZ-UHFFFAOYSA-N
SMILESO=C1C=C(OC2=CC=CC=C12)C1=CC=CC=C1
Molecular Weight (g/mol)222.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=99.0 %
Appearance (Form)Powder
Melting point95°C to 98°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only