Thermo Scientific Chemicals

Fluorescein

Catalog number: 119240250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Fluorescein

Catalog number: 119240250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 119240250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2321-07-5
IUPAC Name
3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Formula
C20H12O5
InChI Key
GNBHRKFJIUUOQI-UHFFFAOYSA-N
SMILES
OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
Appearance (Color)
Red-brown to rust brown to brown
Infrared spectrum
Conforms
Specific absorbance A (1%/1cm)
>=1800 (490 +/- 5 nm , 0.5% aq. soln. of Na2CO3)
Sulfated ash
=<2 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text