2-Fluoroaniline, 99+%, Thermo Scientific Chemicals
2-Fluoroaniline, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoroaniline, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 119275000
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
234.00 
Save 35.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
234.00 
Save 35.35 (15%)
Add to cart
2-Fluoroaniline, 99+%, Thermo Scientific Chemicals
Catalog number119275000
Price (USD)/ Each
198.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
234.00 
Save 35.35 (15%)
-
Add to cart
Chemical Identifiers
CAS348-54-9
IUPAC Name2-fluoroaniline
Molecular FormulaC6H6FN
InChI KeyFTZQXOJYPFINKJ-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless to brown
AppearanceMay darken on storage
Refractive index1.5410 to 1.5430 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only