4-Fluoroaniline, 98%, Thermo Scientific Chemicals
4-Fluoroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoroaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
500 mL
Packaging:
Glass bottle
Catalog number 119291000
Price (USD)/ Each
46.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
54.90 
Save 8.25 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
54.90 
Save 8.25 (15%)
Add to cart
4-Fluoroaniline, 98%, Thermo Scientific Chemicals
Catalog number119291000
Price (USD)/ Each
46.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
54.90 
Save 8.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-fluoroaniline
Molecular FormulaC6H6FN
InChI KeyKRZCOLNOCZKSDF-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1
Molecular Weight (g/mol)111.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to red-brown
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.5385 to 1.5405 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only