Thermo Scientific Chemicals

4-Fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 119295000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

4-Fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 119295000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 119295000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
371-40-4
IUPAC Name
4-fluoroaniline
Molecular Formula
C6H6FN
InChI Key
KRZCOLNOCZKSDF-UHFFFAOYSA-N
SMILES
NC1=CC=C(F)C=C1
Appearance (Color)
Clear light yellow to red-brown
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
Refractive index
1.5385 to 1.5405 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text