5-Fluoro-2-methylaniline, 99%, Thermo Scientific Chemicals
5-Fluoro-2-methylaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-methylaniline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 119461000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-fluoro-2-methylaniline
Molecular FormulaC7H8FN
InChI KeyJLCDTNNLXUMYFQ-UHFFFAOYSA-N
SMILESCC1=CC=C(F)C=C1N
Molecular Weight (g/mol)125.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to purple
GC>=98.5 %
Infrared spectrumConforms
AppearanceMay darken on storage
Appearance (Form)Crystalline solid or liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only