Thermo Scientific Chemicals

3-Fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 119550500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Fluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: 119550500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 119550500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
372-20-3
IUPAC Name
3-fluorophenol
Molecular Formula
C6H5FO
InChI Key
SJTBRFHBXDZMPS-UHFFFAOYSA-N
SMILES
OC1=CC=CC(F)=C1
Appearance
May darken on storage
Appearance (Color)
Clear colorless to yellow-brown
Refractive index
1.5125 to 1.5155 (20°C, 589 nm)
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text