3-Fluorophenol, 98%, Thermo Scientific Chemicals
3-Fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
200 mL
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 119552000
Price (USD)/ Each
539.00
-
Add to cart
Quantity:
200 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
539.00
Add to cart
3-Fluorophenol, 98%, Thermo Scientific Chemicals
Catalog number119552000
Price (USD)/ Each
539.00
-
Add to cart
Chemical Identifiers
CAS372-20-3
IUPAC Name3-fluorophenol
Molecular FormulaC6H5FO
InChI KeySJTBRFHBXDZMPS-UHFFFAOYSA-N
SMILESOC1=CC=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Color)Clear colorless to yellow-brown
Refractive index1.5125 to 1.5155 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only