Thermo Scientific Chemicals

4-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 119571000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 119571000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 119571000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
405-50-5
IUPAC Name
2-(4-fluorophenyl)acetic acid
Molecular Formula
C8H7FO2
InChI Key
MGKPFALCNDRSQD-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(F)C=C1
Appearance (Form)
Crystalline powder or crystals and/or chunks
HPLC
>=97.5 %
Melting point
81°C to 85°C
Infrared spectrum
Conforms
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text