4-Fluorophenyl isocyanate, 99%, Thermo Scientific Chemicals
4-Fluorophenyl isocyanate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorophenyl isocyanate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 119600050
Price (USD)/ Each
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1195-45-5
IUPAC Name1-fluoro-4-isocyanatobenzene
Molecular FormulaC7H4FNO
InChI KeyDSVGFKBFFICWLZ-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.5130 to 1.5150 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only