4-Fluorothiophenol, 97%, Thermo Scientific Chemicals
4-Fluorothiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorothiophenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
View all
Glass bottle
Catalog number 119650100
Price (USD)/ Each
80.80
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
80.80
Add to cart
4-Fluorothiophenol, 97%, Thermo Scientific Chemicals
Catalog number119650100
Price (USD)/ Each
80.80
-
Add to cart
Chemical Identifiers
CAS371-42-6
IUPAC Name4-fluorobenzene-1-thiol
Molecular FormulaC6H5FS
InChI KeyOKIHXNKYYGUVTE-UHFFFAOYSA-N
SMILESFC1=CC=C(S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5490 to 1.5520 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only