Thermo Scientific Chemicals

4-Fluorothiophenol, 97%, Thermo Scientific Chemicals

Catalog number: 119650500
50 g, Glass Bottle, Each
Thermo Scientific Chemicals

4-Fluorothiophenol, 97%, Thermo Scientific Chemicals

Catalog number: 119650500
50 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 119650500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
371-42-6
IUPAC Name
4-fluorobenzene-1-thiol
Molecular Formula
C6H5FS
InChI Key
OKIHXNKYYGUVTE-UHFFFAOYSA-N
SMILES
FC1=CC=C(S)C=C1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Refractive index
1.5490 to 1.5520 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text