Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Thermo Scientific Chemicals

Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Plastic bottle
Catalog number 119690010
Price (USD)
51.10
Each
In stock
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
51.10
Each
Add to cart
Chemical Identifiers
CAS50-00-0
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer5 to 15 % methanol
Color scale=<10 APHA
Free acid (HCOOH)=<0.1 %
Titration35.0 to 39.0 %
Appearance (Form)Clear solution
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; that can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • It May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470890Certificate of AnalysisMay 17, 2025119690050, 119690025, 119690250, 119690010
A0466650Certificate of AnalysisMay 17, 2025119690050, 119690025, 119690250, 119690010
A0462172Certificate of AnalysisJan 30, 2025119690050, 119690025, 119690250, 119690010
A0466650Certificate of AnalysisOct 10, 2024119690050, 119690025, 119690250, 119690010
A0462172Certificate of AnalysisApr 24, 2024119690050, 119690025, 119690250, 119690010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection