Thermo Scientific Chemicals

Formamidinesulfinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 119721000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Formamidinesulfinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 119721000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 119721000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1758-73-2
IUPAC Name
carbamimidoylsulfinic acid
Molecular Formula
CH4N2O2S
InChI Key
FYOWZTWVYZOZSI-UHFFFAOYNA-N
SMILES
NC(=N)S(O)=O
Appearance (Form)
Crystalline powder or crystals
Water
=<0.5 %
Appearance (Color)
White to light yellow
Titration with NaOH
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text