Thermo Scientific Chemicals

Fumaric acid, 99+%, Thermo Scientific Chemicals

Catalog number: 119750100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Fumaric acid, 99+%, Thermo Scientific Chemicals

Catalog number: 119750100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 119750100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
110-17-8
IUPAC Name
(2E)-but-2-enedioic acid
Molecular Formula
C4H4O4
InChI Key
VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES
OC(=O)\C=C\C(O)=O
Appearance (Color)
White
HPLC
>=99.0 %
Appearance (Form)
Crystalline powder
Melting point
295°C to 300°C
Water
=<0.25 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text