Fumaric acid, 99+%
Fumaric acid, 99+%
Fumaric acid, 99+%
Thermo Scientific Chemicals

Fumaric acid, 99+%

CAS: 110-17-8 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 119751000
Price (EUR)
20,15
Online Exclusive
24,30
Save 4,15 (17%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
20,15
Online Exclusive
24,30
Save 4,15 (17%)
Each
Add to cart
Chemical Identifiers
CAS110-17-8
IUPAC Name(2E)-but-2-enedioic acid
Molecular FormulaC4H4O4
InChI KeyVZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILESOC(=O)\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
HPLC>=99.0 %
Appearance (Form)Crystalline powder
Melting point295°C to 300°C
Water=<0.25 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link