Thermo Scientific Chemicals

Furfuryl Alcohol, 98%, Thermo Scientific Chemicals

Catalog number: 119790010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Furfuryl Alcohol, 98%, Thermo Scientific Chemicals

Catalog number: 119790010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 119790010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-00-0
IUPAC Name
(furan-2-yl)methanol
Molecular Formula
C5H6O2
InChI Key
XPFVYQJUAUNWIW-UHFFFAOYSA-N
SMILES
OCC1=CC=CO1
Infrared spectrum
Conforms
Water
=<0.3 %
Refractive index
1.4850 to 1.4880 (20°C, 589 nm)
Appearance (Color)
Clear yellow
Appearance
May darken on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text