Thermo Scientific Chemicals

Furfurylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 119800010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Furfurylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 119800010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 119800010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-89-0
IUPAC Name
1-(furan-2-yl)methanamine
Molecular Formula
C5H7NO
InChI Key
DDRPCXLAQZKBJP-UHFFFAOYSA-N
SMILES
NCC1=CC=CO1
Infrared spectrum
Conforms
Refractive index
1.4890 to 1.4910 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow-brown
GC
>=99.0 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text