Thermo Scientific Chemicals

Furfuryl mercaptan, 97%, Thermo Scientific Chemicals

Catalog number: 119810250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Furfuryl mercaptan, 97%, Thermo Scientific Chemicals

Catalog number: 119810250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 119810250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
98-02-2
IUPAC Name
(furan-2-yl)methanethiol
Molecular Formula
C5H6OS
InChI Key
ZFFTZDQKIXPDAF-UHFFFAOYSA-N
SMILES
SCC1=CC=CO1
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.5290 to 1.5330 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text