Thermo Scientific Chemicals

2-Furoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 119821000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Furoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 119821000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 119821000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-14-2
IUPAC Name
furan-2-carboxylic acid
Molecular Formula
C5H4O3
InChI Key
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CO1
Appearance (Color)
Off-white to beige
Titration with NaOH
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
129°C to 133°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text