Gibberellic Acid, 90%, Thermo Scientific Chemicals
Gibberellic Acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gibberellic Acid, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 119860050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS77-06-5
IUPAC Name(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
Molecular FormulaC19H22O6
InChI KeyIXORZMNAPKEEDV-QTWFBFKQSA-N
SMILESCC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Loss on drying=<3 % (100°C, 7 hrs) (vacuum)
Arsenic (As)=<3 ppm
Infrared spectrumConforms
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only