1x1 image pixel for data collection
Girard's Reagent T, 99%
Girard's Reagent T, 99%
Girard's Reagent T, 99%
Girard's Reagent T, 99%
Thermo Scientific Chemicals

Girard's Reagent T, 99%

CAS: 123-46-6 | C5H14ClN3O | 167.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 119870500
Price (USD)
34.65
Special offer
Online exclusive
Ends: 26-Sep-2025
42.40
Save 7.75 (18%)
Each
In stock
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 26-Sep-2025
42.40
Save 7.75 (18%)
Each
Add to cart
Chemical Identifiers
CAS123-46-6
IUPAC Name[(hydrazinecarbonyl)methyl]trimethylazanium chloride
Molecular FormulaC5H14ClN3O
InChI KeyYSULOORXQBDPCU-UHFFFAOYSA-N
SMILES[Cl-].C[N+](C)(C)CC(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration Argentometric>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point187°C to 192°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection