Thermo Scientific Chemicals

Gluconic Acid, 50 wt. % Solution in Water, Thermo Scientific Chemicals

Catalog number: 119890010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Gluconic Acid, 50 wt. % Solution in Water, Thermo Scientific Chemicals

Catalog number: 119890010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 119890010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
526-95-4
IUPAC Name
potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular Formula
C6H11KO7
InChI Key
HLCFGWHYROZGBI-JJKGCWMISA-M
SMILES
[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Appearance (Color)
Clear yellow to brown-yellow
Titration excess NaOH
49 to 55 %
Appearance (Form)
Solution
Reducing sugars
=<3 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text