Thermo Scientific Chemicals

D-Glucosamine hydrochloride, 98+%

Catalog number: 119900100
10 g, Glass Bottle, Each
Thermo Scientific Chemicals

D-Glucosamine hydrochloride, 98+%

Catalog number: 119900100
10 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 119900100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
66-84-2
IUPAC Name
hydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
Molecular Formula
C6H14ClNO5
InChI Key
CBOJBBMQJBVCMW-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO
Appearance (Form)
Crystalline powder
Titration with HClO4
98.0 to 102.0 % (On dry substance)
Arsenic (As)
=<3 ppm
Specific optical rotation
(after 3 h)
Specific optical rotation
+70.0° to +73.0° (20°C, 589 nm) (c=2.5, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text