Thermo Scientific Chemicals

DL-Glutamic acid monohydrate, 98+%

Catalog number: 119930250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

DL-Glutamic acid monohydrate, 98+%

Catalog number: 119930250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 119930250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19285-83-7
IUPAC Name
2-aminopentanedioic acid hydrate
Molecular Formula
C5H11NO5
InChI Key
OZDAOHVKBFBBMZ-UHFFFAOYNA-N
SMILES
O.NC(CCC(O)=O)C(O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with NaOH
>=98.0 %
Appearance (Form)
Powder
Water
=<15.5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text