Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

L(+)-Glutamic acid monosodium salt monohydrate, 99%

Catalog number: 119940100
10 kg, Plastic Drum, Each
Thermo Scientific Chemicals

L(+)-Glutamic acid monosodium salt monohydrate, 99%

Catalog number: 119940100
10 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 119940100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6106-04-3
IUPAC Name
sodium (4S)-4-amino-4-carboxybutanoate hydrate
Molecular Formula
C5H10NNaO5
InChI Key
GJBHGUUFMNITCI-QTNFYWBSSA-M
SMILES
O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O
Infrared spectrum
Conforms
Loss on drying
=<0.5 % (100°C, 5 hrs)
Specific optical rotation
+24.5° to +25.5° (20°C, 589 nm) (c=10, 2 N HCl)
Appearance (Form)
Crystalline powder or crystals
Arsenic (As)
=<3 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text