L(+)-Glutamine, 99%
L(+)-Glutamine, 99%
L(+)-Glutamine, 99%
Thermo Scientific Chemicals

L(+)-Glutamine, 99%

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic Bottle
Catalog number 119955000
Price (USD)
270.65
Online Exclusive
301.00
Save 30.35 (10%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
270.65
Online Exclusive
301.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS56-85-9
IUPAC Name(2S)-2-amino-4-carbamoylbutanoic acid
Molecular FormulaC5H10N2O3
InChI KeyZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILESN[C@@H](CCC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on drying=<0.3 % (105°C, 3 h)
Sulfated ash=<0.3 %
Iron (Fe)=<30 ppm
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection