Glutaric acid, 99%
Glutaric acid, 99%
Glutaric acid, 99%
Thermo Scientific Chemicals

Glutaric acid, 99%

CAS: 110-94-1 | C5H8O4 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 119960250
Price (USD)
30.65
Online Exclusive
33.40
Save 2.75 (8%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
33.40
Save 2.75 (8%)
Each
Add to cart
Chemical Identifiers
CAS110-94-1
IUPAC Namepentanedioic acid
Molecular FormulaC5H8O4
InChI KeyJFCQEDHGNNZCLN-UHFFFAOYSA-N
SMILESOC(=O)CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Titration with NaOH98.5 to 101.5 %
Melting point95°C to 99°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link