Thermo Scientific Chemicals

Glutaric acid, 99%, Thermo Scientific Chemicals

Catalog number: 119965000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Glutaric acid, 99%, Thermo Scientific Chemicals

Catalog number: 119965000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 119965000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
110-94-1
IUPAC Name
pentanedioic acid
Molecular Formula
C5H8O4
InChI Key
JFCQEDHGNNZCLN-UHFFFAOYSA-N
SMILES
OC(=O)CCCC(O)=O
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
98.5 to 101.5 %
Melting point
95°C to 99°C
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text