Thermo Scientific Chemicals

Glutaric anhydride, 95%, Thermo Scientific Chemicals

Catalog number: 119971000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Glutaric anhydride, 95%, Thermo Scientific Chemicals

Catalog number: 119971000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 119971000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
108-55-4
IUPAC Name
oxane-2,6-dione
Molecular Formula
C5H6O3
InChI Key
VANNPISTIUFMLH-UHFFFAOYSA-N
SMILES
O=C1CCCC(=O)O1
Appearance (Form)
Powder or crystals and/or chunks
GC
>=94 %
Infrared spectrum
Conforms
Appearance (Color)
White to yellow to light beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text