Glutaric anhydride, 95%
Glutaric anhydride, 95%
Glutaric anhydride, 95%
Glutaric anhydride, 95%
Thermo Scientific Chemicals

Glutaric anhydride, 95%

CAS: 108-55-4 | C5H6O3 | 114.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 119975000
Price (USD)
215.65
Special offer
Online exclusive
254.00
Save 38.35 (15%)
Each
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
215.65
Special offer
Online exclusive
254.00
Save 38.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS108-55-4
IUPAC Nameoxane-2,6-dione
Molecular FormulaC5H6O3
InChI KeyVANNPISTIUFMLH-UHFFFAOYSA-N
SMILESO=C1CCCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
GC>=94 %
Infrared spectrumConforms
Appearance (Color)White to yellow to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection