Thermo Scientific Chemicals

Glutaryl dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 119990250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Glutaryl dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 119990250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 119990250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2873-74-7
IUPAC Name
pentanedioyl dichloride
Molecular Formula
C5H6Cl2O2
InChI Key
YVOFTMXWTWHRBH-UHFFFAOYSA-N
SMILES
ClC(=O)CCCC(Cl)=O
Appearance (Form)
Liquid
Titration RCOCl
>=96.0 %
Refractive index
1.4720 to 1.4760 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow to red-brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text