eBioscience™ Streptavidin PE Conjugate
Product Image
Invitrogen™

eBioscience™ Streptavidin PE Conjugate

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. StreptavidinRead more
Have Questions?
Catalog NumberQuantity
12-4317-87250 μg
Catalog number 12-4317-87
Price (USD)
87.65
Online Exclusive
117.00
Save 29.35 (25%)
Each
In stock
Add to cart
Quantity:
250 μg
Price (USD)
87.65
Online Exclusive
117.00
Save 29.35 (25%)
Each
Add to cart
The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser
Blue Laser, Green Laser, Yellow-Green Laser

Emit
578 nm

Excite
488 - 561 nm

Reported Application
Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.2 mg/mL
Label or DyePE (Phycoerythrin)
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity250 μg
Target MoleculeBiotin
ConjugatePE (Phycoerythrin)
FormLiquid
Product LineeBioscience™
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection