Nalgene™ Disposable Polypropylene Robotic Reservoirs
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Thermo Scientific™

Nalgene™ Disposable Polypropylene Robotic Reservoirs

Increase walk-away time and assay efficiency with these disposable polypropylene robotic reservoirs, available as a standard microplate format with 300mL volume.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeSterility
1200-1300FlatNon-sterile
1200-1301FlatSterile
1200-2300ConvolutedNon-sterile
1200-2301ConvolutedSterile
Catalog number 1200-1300
Price (USD)
325.65
Online Exclusive
344.00
Save 18.35 (5%)
Case of 40
Add to cart
Bottom Shape:
Flat
Sterility:
Non-sterile
Price (USD)
325.65
Online Exclusive
344.00
Save 18.35 (5%)
Case of 40
Add to cart

Increase walk-away time and assay efficiency with Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs. Featuring a standard microplate format with 300mL volume, the convoluted design minimizes dead volume and has baffles to reduce splashing. The flat bottom version is designed to meet a variety of assay needs in both manual and automated environments.

  • Convoluted bottom design is most effective with a 96-well format.
  • Flat Bottom design can be used with single, six-, 12-, 96-, 384- and 1536-well formats.
  • Polypropylene construction withstands robotic liquid handling, making it easier to run automated assays and increasing walk-away time.
  • Reservoir is compatible with most robotic arms, plate stackers (hotels), plate cranes, and platform configurations.
  • Reservoirs are RNase- & DNase-free.
  • Reservoir volume is 300mL (345mL brim capacity), with molded-in graduations every 100mL.
  • Reservoir is nestable for easy automated stacking and handling.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceMeets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
ColorNatural
Dimensions (L x W x H)3.4 x 5 x 1.75 in. (86 x 128 x 43 mm)
Height (English)1.75 in.
Height (Metric)43 mm
Length (English)3.4 in.
Length (Metric)86 mm
No. per Case40
No. per Pack4
Plate CompatibilitySingle, 6-Well Plates, 12-Well Plates, 96-Well Plates, 384-Well Plates, 1536-Well Plates
Width (English)5 in.
Width (Metric)128 mm
Bottom ShapeFlat
Capacity (Metric)300 mL
MaterialPolypropylene
SterilityNon-sterile
TypeReservoir
Unit SizeCase of 40
nalgene-certificate-of-compliance

Certificates of Compliance

Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Nalgene and Nunc products.
Start your request  

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link