Thermo Scientific Chemicals

Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals

Catalog number: 120000010
1 g, Each
Thermo Scientific Chemicals

Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals

Catalog number: 120000010
1 g, Each
Quantity
Catalog number: 120000010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
70-18-8
IUPAC Name
2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
Molecular Formula
C10H17N3O6S
InChI Key
RWSXRVCMGQZWBV-UHFFFAOYNA-N
SMILES
NC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O
Appearance (Form)
Crystalline powder or crystals
Iron (Fe)
=<10 ppm
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Titration Iodimetric
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text