Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals
Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
100 g
500 g
Catalog number 120000050
Price (USD)/ Each
56.65
Online exclusive
63.20 
Save 6.55 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
63.20 
Save 6.55 (10%)
Add to cart
Glutathione, 98%, for analysis, reduced, Thermo Scientific Chemicals
Catalog number120000050
Price (USD)/ Each
56.65
Online exclusive
63.20 
Save 6.55 (10%)
-
Add to cart
Chemical Identifiers
CAS70-18-8
IUPAC Name2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
Molecular FormulaC10H17N3O6S
InChI KeyRWSXRVCMGQZWBV-UHFFFAOYNA-N
SMILESNC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Iron (Fe)=<10 ppm
Appearance (Color)White to almost white
Infrared spectrumConforms
Titration Iodimetric97.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only