Thermo Scientific Chemicals

Glycine anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 120081000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Glycine anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 120081000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 120081000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
106-57-0
IUPAC Name
piperazine-2,5-dione
Molecular Formula
C4H6N2O2
InChI Key
BXRNXXXXHLBUKK-UHFFFAOYSA-N
SMILES
O=C1CNC(=O)CN1
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Loss on drying
=<0.5 % (105°C, 4 h)
Appearance (Color)
White to light yellow to beige
Assay
>=97.5 % (ex N)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text