Glycine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Glycine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 120090025
Price (USD)/ Each
304.00
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
304.00
Add to cart
Glycine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Catalog number120090025
Price (USD)/ Each
304.00
-
Add to cart
Chemical Identifiers
CAS623-33-6
IUPAC Name2-ethoxy-2-oxoethan-1-aminium chloride
Molecular FormulaC4H10ClNO2
InChI KeyTXTWXQXDMWILOF-UHFFFAOYSA-N
SMILES[Cl-].CCOC(=O)C[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Free acid (HCl)=<0.4 %
Appearance (Color)White
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only