Thermo Scientific Chemicals

Glycine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 120095000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Glycine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 120095000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 120095000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
623-33-6
IUPAC Name
2-ethoxy-2-oxoethan-1-aminium chloride
Molecular Formula
C4H10ClNO2
InChI Key
TXTWXQXDMWILOF-UHFFFAOYSA-N
SMILES
[Cl-].CCOC(=O)C[NH3+]
Infrared spectrum
Conforms
Titration with HClO4
>=98.5 %
Free acid (HCl)
=<0.4 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text