Glycoluril, 97%, Thermo Scientific Chemicals
Glycoluril, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycoluril, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 120135000
Price (USD)/ Each
249.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
294.00 
Save 44.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
249.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
294.00 
Save 44.35 (15%)
Add to cart
Glycoluril, 97%, Thermo Scientific Chemicals
Catalog number120135000
Price (USD)/ Each
249.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
294.00 
Save 44.35 (15%)
-
Add to cart
Chemical Identifiers
CAS496-46-8
IUPAC Nameoctahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
Molecular FormulaC4H6N4O2
InChI KeyVPVSTMAPERLKKM-UHFFFAOYSA-N
SMILESO=C1NC2NC(=O)NC2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Assay>=96.0 % (N)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only