Thermo Scientific Chemicals

Glycylglycine, 99+%

Catalog number: 120140250
25 g, Each
Thermo Scientific Chemicals

Glycylglycine, 99+%

Catalog number: 120140250
25 g, Each
Quantity
Catalog number: 120140250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
556-50-3
IUPAC Name
hydrogen 2-(2-aminoacetamido)acetic acid chloride
Molecular Formula
C4H9ClN2O3
InChI Key
YHBAZQDEMYQPJL-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCC(=O)NCC(O)=O
Appearance (Color)
White
Heavy metals (as Pb)
=<5 ppm
Infrared spectrum
Conforms
Loss on drying
=<0.5 % (105°C, 4 h)
pH
5.5 to 6.5 (5 % in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text