Thermo Scientific Chemicals

18-β-Glycyrrhetinic Acid, 98+%, Thermo Scientific Chemicals

Catalog number: 120150050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

18-β-Glycyrrhetinic Acid, 98+%, Thermo Scientific Chemicals

Catalog number: 120150050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 120150050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-53-4
IUPAC Name
(2S,4aS,6aS,6bR,8aR,10S,12aS,12bR,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylic acid
Molecular Formula
C30H46O4
InChI Key
MPDGHEJMBKOTSU-YKLVYJNSSA-N
SMILES
CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O
Appearance (Form)
Crystalline powder
Specific optical rotation
on dried substance
Appearance (Color)
White to off-white
Titration with NaOH
98.0 to 102.0 % (on dried substance)
Sulfated ash
=<0.2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text