Thermo Scientific Chemicals

Glyoxylic acid monohydrate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 120175000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Glyoxylic acid monohydrate, 98%, pure, Thermo Scientific Chemicals

Catalog number: 120175000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 120175000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
563-96-2
IUPAC Name
2,2-dihydroxyacetate
Molecular Formula
C2H3O4
InChI Key
GOCCREQJUBABAL-UHFFFAOYSA-M
SMILES
OC(O)C([O-])=O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder and/or chunks
Melting point
48°C to 52°C
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text