Guaiacol, 99+%
Guaiacol, 99+%
Guaiacol, 99+%
Thermo Scientific Chemicals

Guaiacol, 99+%

CAS: 90-05-1 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Glass bottle
Catalog number 120190010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS90-05-1
IUPAC Name2-methoxyphenol
Molecular FormulaC7H8O2
InChI KeyLHGVFZTZFXWLCP-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid or crystalline mass
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.5420 to 1.5470 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link